Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p57 Kip2 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524428

Catalog No. PIPA524428

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »


Description

Description

This gene is imprinted, with preferential expression of the maternal allele. The encoded protein is a tight-binding, strong inhibitor of several G1 cyclin/Cdk complexes and a negative regulator of cell proliferation. Mutations in this gene are implicated in sporadic cancers and Beckwith-Wiedemann syndorome, suggesting that this gene is a tumor suppressor candidate. Three transcript variants encoding two different isoforms have been found for this gene.
Specifications

Specifications

p57 Kip2
Polyclonal
PBS with 0.09% sodium azide
P49918, P49919
BWCR, BWS, KIP2, WBS, p57, cyclin-dependent kinase inhibitor 1C, cyclin-dependent kinase inhibitor p57, p57Kip2
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
1028, 12577
Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
CDKN1C
Liquid
CDKN1C
KLH conjugated synthetic peptide between 34-64 amino acids from the N-terminal region of human CDKN1C
Protein A, Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit