Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CDKN2B Monoclonal Antibody (DCS114.1), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA112294
Description
CDKN2B Monoclonal antibody specifically detects CDKN2B in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.Specifications
CDKN2B | |
Monoclonal | |
1 mg/mL | |
PBS with 0.08% sodium azide | |
P42772 | |
CDKN2B | |
His-tagged recombinant full length p15 protein. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
DCS114.1 | |
Unconjugated | |
CDKN2B | |
AV083695; CDK inhibitory protein; CDK4B inhibitor; CDK4I; Cdkn2b; cyclin dependent kinase inhibitor 2B; Cyclin dependent kinase inhibitor 2B (p15, inhibits CDK4); cyclin-dependent kinase 4 inhibitor B; cyclin-dependent kinase inhibitor 2B (p15, inhibits CDK4); cyclin-dependent kinase inhibitor p15; cyclin-dependent kinase inhibitor p15INK4b; cyclin-dependent kinase inhibitor protein; cyclin-dependent kinases 4 and 6 binding protein; Ink4; INK4B; MTS2; MTS-2; Multiple tumor suppressor 2; p14_CDK inhibitor; p14_INK4B; p14-INK4b; P15; p15 CDK inhibitor; p15(INK4b); p15_INK4B; p15INK4b; p15-INK4b; TP15 | |
Mouse | |
Ammonium sulfate precipitation | |
RUO | |
1030 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction