Learn More
CDKN2C, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human CDKN2C peptide using ARM Technology.
Manufacturer:  Abnova Corporation H00001031K
Catalog No. 89-200-217
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
Description
The protein encoded by this gene is a member of the INK4 family of cyclin-dependent kinase inhibitors. This protein has been shown to interact with CDK4 or CDK6, and prevent the activation of the CDK kinases, thus function as a cell growth regulator that controls cell cycle G1 progression. Ectopic expression of this gene was shown to suppress the growth of human cells in a manner that appears to correlate with the presence of a wild-type RB1 function. Studies in the knockout mice suggested the roles of this gene in regulating spermatogenesis, as well as in suppressing tumorigenesis. Two alternatively spliced transcript variants of this gene, which encode an identical protein, have been reported. [provided by RefSeq
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)
Expression: Overexpression vector and transfection into 293H cell line
Order Info
1. Customer may provide cell or tissue lysate for antibody screening.
2. Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
Specifications
CDKN2C
1mg
cyclin-dependent kinase inhibitor 2C (p18, inhibits CDK4)
CDKN2C
Liquid
CDKN2C
A synthetic peptide of human CDKN2C
1mg x up to 3 Abs
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
ELISA, Western Blot
Unconjugated
In 1X PBS, pH 7.4
CDKN2C
INK4C, p18, p18-INK4C
Rabbit
IgG
RUO
Primary
1031