Learn More
CDX1 Rabbit anti-Human, Mouse, HRP, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA523060
Catalog No. PIPA523060
Add to cart
Description
Description
The transcription factors Cdx1 and Cdx2 are members of the caudal-related homeobox gene family based on their homology to the factor caudal in Drosophila melanogaster. A wide variety of in vivo and in vitro studies have suggested that Cdx1 and Cdx2 are important in controlling epithelial cell differentiation, proliferation, and function in the intestine.
Specifications
Specifications
Cdx1
Polyclonal
HRP
CDX1
Liquid
CDX1
A synthetic peptide made to an internal region of the human Cdx1 protein (within residues 30-150).
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Immunohistochemistry
0.55 mg/mL
PBS with no preservative
P47902, P18111
caudal type homeo box transcription factor 1, caudal type homeobox transcription factor 1, caudal-type homeobox protein 1, caudal-type homeobox protein CDX1, homeobox protein CDX-1
Rabbit
IgG
100 μL
4° C, store in dark
Primary
1044, 12590