Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDX1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523056

Catalog No. PIPA523056


Add to cart

Description

Description

The transcription factors Cdx1 and Cdx2 are members of the caudal-related homeobox gene family based on their homology to the factor caudal in Drosophila melanogaster. A wide variety of in vivo and in vitro studies have suggested that Cdx1 and Cdx2 are important in controlling epithelial cell differentiation, proliferation, and function in the intestine.
Specifications

Specifications

Cdx1
Polyclonal
Unconjugated
CDX1
Liquid
CDX1
A synthetic peptide made to an internal region of the human Cdx1 protein (within residues 30-150).
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
0.55 mg/mL
PBS with 30% glycerol and 0.05% sodium azide
P47902, P18111
caudal type homeo box transcription factor 1, caudal type homeobox transcription factor 1, caudal-type homeobox protein 1, caudal-type homeobox protein CDX1, homeobox protein CDX-1
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
1044, 12590
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit