Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDX2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA124393

Catalog No. PA124393


Add to cart

Description

Description

PA1-24393 detects CDX2 in Human samples. PA1-24393 has been successfully used in Western Blot procedures. PA1-24393 immunogen corresponds to Polyclonal antibody from rabbits immunized with a synthetic peptide region of human CDX2. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

CDX2 is a member of caudal type homeo box family of transcription factor. CDX-2 is a permissive factor that influences basal CaBP expression in enterocytes; HNF-1alpha; modulates CaBP gene expression during cellular differentiation. CDX2, but not CDX1, interacts with the MUC2 promoter and activates MUC2 transcription, and plays an important role in the differentiation of goblet cells.
Specifications

Specifications

CDX2
Polyclonal
Unconjugated
CDX2
Liquid
CDX2
Synthetic peptide region of human CDX2.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
1 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
Q99626
CDX-3, CDX3
Rabbit
IgG
50 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
1045
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit