Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CEP164 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA524461
Description
This antibody is predicted to react with bovine and rat based on sequence homology.
Plays a role in microtubule organization and/or maintece for the formation of primary cilia (PC), a microtubule-based structure that protrudes from the surface of epithelial cells. Plays a critical role in G2/M checkpoint and nuclear divisions. A key player in the DNA damage-activated ATR/ATM signaling cascade since it is required for the proper phosphorylation of H2AX, RPA,CHK2 and CHK1. Plays a critical role in chromosome segregation, acting as a mediator required for the maintece of genomic stability through modulation of MDC1, RPA and CHK1.Specifications
CEP164 | |
Polyclonal | |
CEP164 | |
centrosomal protein of 164 kDa | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
22897 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Western Blot | |
Unconjugated | |
Q9UPV0 | |
CEP164 | |
KLH conjugated synthetic peptide between 246-274 amino acids from the N-terminal region of human CE164 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction