Learn More
Thermo Scientific Lab Vision CEA (Carcinoembryonic Antigen)/CD66e Ab-2, Rabbit Polyclonal Antibody
Ensure accurate, reproducible results in immunohistochemistry, western blotting and immunoprecipitation experiments.
Manufacturer: Thermo Scientific RB368A
Description
Ensure accurate, reproducible results in immunohistochemistry, western blotting and immunoprecipitation experiments with Thermo Scientific CEA (Carcinoembryonic Antigen)/CD66e Ab-2, Rabbit Polyclonal Antibody.
Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human and Mouse. Weakly reacts with Rat. Does not react with dog. Others not known
Immunogen: Human CEA isolated from hepatic metastasis of colon adenocarcinoma
Molecular Weight: 180 kDa
Positive Control: LS174T cells or colon CA
Cellular Localization: Cytoplasmic and luminal membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunoprecipitation (Native and denatured)
Specifications
CEA (Carcinoembryonic Antigen)/CD66e Ab-2 | |
Polyclonal | |
Unconjugated | |
Concentrated | |
Human CEA isolated from hepatic metastasis of colon adenocarcinoma | |
LS174T cells or colon CA | |
IVD | |
Primary | |
Polyclonal |
Immunoprecipitation, Western Blot | |
Polyclonal | |
Purified Ab with BSA and Azide | |
Rabbit | |
IgG | |
1mL | |
Oncogenesis | |
Primary | |
Human |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.