Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CEACAM1 Mouse anti-Human, Clone: GM8G5, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA117758

Catalog No. 01-670-171


Add to cart

Description

Description

This antibody recognizes native human CEACAM1 protein expressed in transfected mammalian cells.

CEACAM1 (BGP/CD66a) is a transmembrane glycoprotein which belongs to the carcinoembryonic antigen (CEA) gene family. It is expressed on cells of epithelial and myeloid origin and mediates homophilic intercellular interactions that influence cellular growth, immune cell activation, and tissue morphogenesis. CEACAM1 is a putative tumour suppressor based on diminished expression in some aggressive types of cancer cells. The anti-tumour effect may be due to inhibition of tumour angiogenesis, possibly by increased secretion of anti-angiogenic molecules from the cells. Like all members of the CEACAM family, it consists of a single N domain, with structural homology to the immunoglobulin variable domains, followed by two immunoglobulin constant-like A (A1, A2) and one B domain. While the N, A1 and B domains can also be found in other CEA-family members, the A2 domain of CEACAM1 differs from those found in other CEACAM.
Specifications

Specifications

CEACAM1
Monoclonal
2 mg/mL
PBS with no preservative; pH 7.2
P13688
BGP, BGP1, BGPI, CD66a antigen, antigen CD66, carcinoembryonic antigen-related cell adhesion molecule 1
Mouse
IgG1
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
634
ELISA, Flow Cytometry, Immunohistochemistry (Frozen)
GM8G5
Unconjugated
CEACAM1
Liquid
CEACAM1
cDNA encoding the extracellular region of human CEACAM1-A2
Protein G
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit