Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CEACAM7 Monoclonal Antibody (BAC2), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA117764
Description
This antibody recognizes native human CEACAM7 protein expressed in transfected mammalian cells.
CEACAM7 Monoclonal antibody specifically detects CEACAM7 in Human samples. It is validated for ELISA, Flow Cytometry.Specifications
CEACAM7 | |
Monoclonal | |
2 mg/mL | |
PBS with no preservative; pH 7.2 | |
Q14002 | |
CEACAM7 | |
Purified CEACAM7/Fc fusion protein. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Flow Cytometry | |
BAC2 | |
Unconjugated | |
CEACAM7 | |
BGP; BGP7; carcinoembryonic antigen CGM2; carcinoembryonic antigen gene family member 2; carcinoembryonic antigen related cell adhesion molecule 7; carcinoembryonic antigen-related cell adhesion molecule 7; CEA; CEACAM7; CGM2 | |
Mouse | |
Protein G | |
RUO | |
1087 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction