Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CEACAM7, Polyclonal, Invitrogen™

Chicken Polyclonal Antibody

Supplier:  Thermo Scientific PA127881

Catalog No. PIPA127881

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.


Description

Description

PA1-27881 detects CEACAM7 from human samples.PA1-27881 may be successfully used in Western blot applications.The PA1-27887 immunogen is a synthetic peptide corresponding to human Carcinoembryonic antigen-related cell adhesion molecule 7.Store at 4ºC short term. For extended storage aliquot and store at -20ºC or below. Avoid freeze-thaw cycles.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CEACAM7
Polyclonal
CEACAM7
CEA, CGM2
Chicken
Antigen affinity chromatography
RUO
1087
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Purified
Western Blot
Unconjugated
Q14002
CEACAM7
Synthetic peptide corresponding to human Carcinoembryonic antigen-related cell adhesion molecule 7.
50 μg
Primary
Human
Antibody
IgY
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit