Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CEACAM8 Mouse anti-Human, Clone: GM2H6, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA117768

Catalog No. 01-670-181


Add to cart

Description

Description

This antibody recognizes native human CEACAM8 protein expressed in transfected mammalian cells.

CEA-related cell adhesion molecule 8 (CEACAM8, CD66b) belongs to the carcinoembryonic antigen (CEA) gene family. It encodes a glycosylphosphatidylinositol (GPI)-linked glycoprotein with a MW of 95kDa which is expressed in cells of the granulocyte-lineage. It is expressed in neutrophils and eosinophils and is characterized as a granulocyte-specific activation antigen. CEACAM8 could serve as a marker for granulocyte activities. Like all members of the CEA family, it consists of a single N domain, with structural homology to the immunoglobulin variable domains, followed by two immunoglobulin constant-like A and B domains.
Specifications

Specifications

CEACAM8
Monoclonal
2 mg/mL
PBS with no preservative; pH 7.2
P31997
CD66b, CD67, CGM6, NCA-95
Mouse
IgG1
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
1088
ELISA, Flow Cytometry
GM2H6
Unconjugated
CEACAM8
Liquid
CEACAM8
cDNA encoding human CEACAM8
Protein G
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit