Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDK20 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA528208

Catalog No. PIPA528208


Add to cart

Description

Description

PA5-28208 targets Cell cycle-related kinase3 in WB applications and shows reactivity with Human samples. The PA5-28208 immunogen is recombit fragment corresponding to a region within amino acids 7 and 319 of Human Cell cycle-related kinase3. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

The protein encoded by this gene contains a kinase domain most closely related to the cyclin-dependent protein kinases. The encoded kinase activates CDK2 and is involved in cell growth. Alternatively spliced transcript variants encoding distinct isoforms have been reported.
Specifications

Specifications

CDK20
Polyclonal
Unconjugated
CDK20
Liquid
CDK20
Recombit fragment corresponding to a region within amino acids 7 and 319 of Human Cell cycle-related kinase3
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunocytochemistry, Immunofluorescence, Western Blot
0.8 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Q8IZL9
CCRK, CDCH, P42, PNQALRE, CAK-kinase p42, CDK-activating kinase p42, cell cycle-related kinase, cell division protein kinase 20, cyclin-dependent protein kinase H, cyclin-kinase-activating kinase p42
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
23552
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit