Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Betacellulin/BTC Mouse anti-Human, Clone: 34362, R&D Systems™

Mouse Monoclonal Antibody

$119.60 - $839.00

Specifications

Antigen Betacellulin/BTC
Immunogen E. coli-derived recombinant human Betacellulin Asp32-Tyr111 Accession # P35070
Clone 34362
Applications ELISA
Conjugate Unconjugated
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB261SP
View Documents
R&D Systems
MAB261SP
25 ug
Each for $119.60
Add to cart
 
MAB261100
View Documents
R&D Systems
MAB261100
100 ug
Each for $377.00
Add to cart
 
MAB261500
View Documents
R&D Systems
MAB261500
500 ug
Each for $839.00
Add to cart
 
Description

Description

Betacellulin/BTC Monoclonal antibody MAB261 Specifically detects Betacellulin/BTC in Human samples. It is validated for ELISA Capture
Specifications

Specifications

Betacellulin/BTC
34362
Unconjugated
P35070
Monoclonal
Primary
Detects human Betacellulin in ELISAs and Western blots. In Western blots, this antibody does not cross-react with recombinant human (rh) EGF, rhHB-EGF, rhTGF-alpha , rhHRG-alpha , or rhAR under non-reducing conditions.
Protein A or G purified from ascites
E. coli-derived recombinant human Betacellulin Asp32-Tyr111 Accession # P35070
ELISA
Purified
Mouse
IgG1
685
RUO
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit