Learn More
CENP-E Mouse anti-Human, Clone: 177, MilliporeSigma™
Mouse Monoclonal Antibody
Manufacturer:  MilliporeSigma 051502
Catalog No. 05-150-2MI
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Use Anti-CENP-E Antibody, clone177 (Mouse Monoclonal Antibody) validated in immunocytochemistry, immunohistochemistry, western blot, immunoprecipitation to detect CENP-E also known as centromere protein E (312kD).
Specifications
Specifications
CENP-E
Monoclonal
Unconjugated
NP_001804
Mouse
Protein G purfied
Primary
2-8°C for one year
IgG1 κ
Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
177
Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide.
CENP-E3, CENP-E
Native protein corresponding to human CENP-E.
100 μg
Human
Purified