Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CENPC Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523651

Catalog No. PIPA523651


Add to cart

Description

Description

Centromere protein C 1 is a centromere autoantigen and a component of the inner kinetochore plate. The protein is required for maintaining proper kinetochore size and a timely transition to anaphase. A putative pseudogene exists on chromosome 12.
Specifications

Specifications

CENPC
Polyclonal
PBS with 0.09% sodium azide
Q03188
CENP-C, CENPC, MIF2, hcp-4, CENP-C 1, centromere autoantigen C, centromere autoantigen C1, interphase centromere complex protein 7
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
1060
Western Blot
Unconjugated
CENPC
Liquid
CENPC
KLH conjugated synthetic peptide between 441-467 amino acids from the central region of human CENPC1
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit