Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CENPI Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA526624
Description
CENPI is involved in the response of gonadal tissues to follicle-stimulating hormone.Specifications
CENPI | |
Polyclonal | |
CENPI | |
CENP-I, FSHPRH1, LRPR1, Mis6, FSH primary response 1, FSH primary response protein 1, follicle-stimulating hormone primary response protein, interphase centromere complex protein 19, leucine-rich primary response protein 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2491 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
Unconjugated | |
Q92674 | |
CENPI | |
KLH conjugated synthetic peptide between 388-414 amino acids from the central region of human CENPI | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction