Learn More
CENPP Mouse anti-Human, Clone: 3G8, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA523311
Catalog No. PIMA523311
Add to cart
Description
Description
Peptide Sequence: MDAELAEVRA LQAEIAALRR ACEDPPAPWE EKSRVQKSFQ AIHQFNLEGW KSSKDLKNQL GHLESELSFL STLTGINIRN HSKQTEDLTS TEMTEKSI
CENPP is a subunit of a CENPH (MIM 605607)-CENPI (MIM 300065)-associated centromeric complex that targets CENPA (MIM 117139) to centromeres and is required for proper kinetochore function and mitotic progression (Okada et al., 2006 [PubMed 16622420]).
Specifications
Specifications
CENPP
Monoclonal
0.2-1.0 mg/mL
PBS with no preservative; pH 7.4
Q6IPU0
CENP-P
Mouse
IgG2a, kappa
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
401541
ELISA, Western Blot
3G8
Unconjugated
CENPP
Liquid
CENPP
CENPP (NP_001012267, 1 a.a. ∽ 98 a.a) partial recombit protein with GST tag. MW of the GST tag alone is 26 KDa.
Affinity chromatography
RUO
Antibody
Monoclonal
Human