Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ArfGAP with coiled-coil, ankyrin repeat and PH domains 2, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human CENTB2 protein.

Manufacturer:  Abnova Corporation H00023527B01

Catalog No. 89-007-287


Add to cart

Description

Description

Sequence: MKMTVDFEECLKDSPRFRAALEEVEGDVAELELKLDKLVKLCIAMIDTGKAFCVANKQFMNGIRDLAQYSSNDAVVETSLTKFSDSLQEMINFHTILFDQTQRSIKAQLQNFVKEDLRKFKDAKKQFEKVSEEKENALVKNAQVQRNKQHEVEEATNILTATRKCFRHIALDYVLQINVLQSKRRSEILKSMLSFMYAHLAFFHQGYDLFSELGPYMKDLGAQLDRLVVDAAKEKREMEQKHSTIQQKDFSSDDSKLEYNVDAANGIVMEGYLFKRASNAFKTWNRRWFSIQNNQLVYQKKFKDNPTVVVEDLRLCTVKHCEDIERRFCFEVVSPTKSCMLQADSEKLRQAWIKAVQTSIATAYREKGDESEKLDKKSSPSTGSLDSGNESKEKLLKGESALQRVQCIPGNASCCDCGLADPRWASINLGITLCIECSGIHRSLGVHFSKVRSLTLDTWEPELLKLMCELGNDVINRVYEANVEKMGIKKPQPGQRQEKEAYIRAKYVERKFVDKYSISLSPPEQQKKFVSKSSEEKRLSISKFGPGDQVRASAQSSVRSNDSEIQQSSDDGRESLPSTVSANSLYEPEGERQDSSMFLDSKHLNPGLQLYRASYEKNLPKMAEALAHGADVNWANSEENKATPLIQAVLGGSLVTCEFLLQNGANVNQRDVQGRGPLHHATVLGHTGQVCLFLKRGANQHATDEEGKDPLSIAVEAANADIVTLLRLARMNEEMRESEGLYGQPGDETYQDIFRDFSQMASNNPEKLNRFQQDSQKF
Specifications

Specifications

ArfGAP with coiled-coil, ankyrin repeat and PH domains 2
Unconjugated
No additive
NM_012287.3
CENTB2|CNT-B2|KIAA0041
Mouse
50 uL
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
23527
Western Blot
Mouse polyclonal antibody raised against a full-length human CENTB2 protein.
ACAP2
Serum
ACAP2
CENTB2 (NP_036419.2, 1 a.a. ~ 778 a.a) full-length human protein.
RUO
Y
Polyclonal
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit