Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Centrin 1 Rabbit anti-Human, Mouse, Xenopus laevis, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA125039

Catalog No. PA125039


Add to cart

Description

Description

PA1-25039 detects Centrin 1 from human samples. PA1-25039 is expected to cross react with mouse, Xenopus laevis, and Drosophilia melanogaster due to sequence homology. PA1-25039 has been successfully used in Western blot and IF/ICC procedures. The PA1-25039 immunogen is a synthetic peptide, conjugated to KLH, corresponding to residues 157-172 of human Centrin 1, with an N-terminal added lysine. This sequence is highly conserved in centrin 1 of many species, e.g. mouse, Drosophila, and Xenopus. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

The protein encoded by this gene plays important roles in the determination of centrosome position and segregation, and in the process of microtubule severing. This encoded protein is localized to the centrosome of interphase cells, and redistributes to the region of the spindle poles during mitosis, reflecting the dynamic behavior of the centrosome during the cell cycle.
Specifications

Specifications

Centrin 1
Polyclonal
Unconjugated
CETN1
Liquid
CETN1, cetn1.S
Synthetic peptide, conjugated to KLH, corresponding to residues 157-172 of human Centrin 1, with an N-terminal added lysine. This sequence is highly c
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Xenopus laevis
Immunocytochemistry, Immunofluorescence, Western Blot
0.70 mg/mL
0.01M PBS with 1% BSA and 15mM sodium azide; pH 7.4
Q12798, P41209
CEN1, CETN
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
1068, 26369, 379819
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit