Learn More
Cep63 Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Manufacturer:  MilliporeSigma 061292
Catalog No. 06-129-2MI
Add to cart
Description
Description
Use Anti-Cep63 Antibody (Rabbit Polyclonal Antibody) validated in western blot, immunocytochemistry to detect Cep63 also known as Centrosomal protein of 63 kDa.
Specifications
Specifications
Cep63
Unconjugated
Affinity Purified
Rabbit
Affinity purified
2-8°C for 12 months
Polyclonal
Human, Murine
Immunocytochemistry, Western Blot
Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
CEP63
Recombinant protein corresponding to human Cep63.
100μL
Primary
NP_001035842.1