Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CEP63 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524734

Catalog No. PIPA524734

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

CEP63 is a protein with six coiled-coil domains. The protein is localized to the centrosome, a non-membraneous organelle that functions as the major microtubule-organizing center in animal cells.
Specifications

Specifications

CEP63
Polyclonal
PBS with 0.09% sodium azide
Q96MT8
centrosomal protein of 63 kDa, centrosome protein CEP63
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
80254
Immunohistochemistry, Western Blot
Unconjugated
CEP63
Liquid
CEP63
KLH conjugated synthetic peptide between 520-548 amino acids from the central region of human CEP63
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit