Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-CEP68, Polyclonal, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP140351

Catalog No. NBP140351

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

CEP68
1.0 mg/mL
Western Blot 1:2000-1:10000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 5-15 ug/mg lysate, Immunohistochemistry-Paraffin 1:10-1:500
Affinity Purified
CEP68
The immunogen for this product maps to a region between residue 350 and 400 of human centrosomal protein 68kDa using the numbering given in entry NP_055962.2 (GeneID 23177).
Immunogen affinity purified
RUO
Primary
23177
Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Unconjugated
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
centrosomal protein 68kDa, centrosomal protein of 68 kDa, Cep68, FLJ36750, KIAA0582FLJ25920
Rabbit
IgG
100 ul
Store at 4C. Do not freeze.
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only