Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BITE Rabbit anti-Hamster, Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524143

Catalog No. PIPA524143


Add to cart

Description

Description

This antibody is predicted to react with bovine and rat based on sequence homology.

Specifications

Specifications

BITE
Polyclonal
PBS with 0.09% sodium azide
Q8NHQ1
BITE, centrosomal protein of 70 kDa, p10-binding protein
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
100767664, 80321
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
CEP70
Liquid
CEP70
KLH conjugated synthetic peptide between 309-338 amino acids from the central region of human CEP70
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Hamster, Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit