Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CEPT1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523876

Catalog No. PIPA523876


Add to cart

Description

Description

This antibody is predicted to react with mouse, rat and Xenopus based on sequence homology.

Choline phosphotransferase catalyses the final step in thesynthesis of phosphatidylcholine by the transfer of phosphocholine from CDP-choline to diacylglycerol. The synthesis ofphosphatidyl ethanolamine by ethanolamine phosphotransferase occurs using an analogous reaction. This gene codes for acholine/ethanolamine phosphotransferase. The protein can synthesize either choline- or ethanolamine- containing phospholipids. Two alternatively spliced transcripts encoding the same isoform have been identified.
Specifications

Specifications

CEPT1
Polyclonal
Unconjugated
CEPT1
Liquid
CEPT1
KLH conjugated synthetic peptide between 29-57 amino acids from the N-terminal region of human CEPT1
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide
Q9Y6K0
choline/ethanolaminephosphotransferase, choline/ethanolaminephosphotransferase 1, hCEPT1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
10390
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit