Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERK Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA112768

Catalog No. 01-675-788


Add to cart

Description

Description

A suggested positive control for this product is heart, brain, skeletal muscle, kidney or liver lysate.

Ceramide kinase (CERK) is a novel lipid kinase. It is responsible for catalyzing the phosphorylation that produces a signaling lipid called ceramide-1-phosphate (C1P). C1P is an important biological mediator that induces inflammatory pathways such as those blocked by many allergy medications. Findings suggest that CERK could be a novel target for a new generation of anti-inflammatory/anti-trauma therapeutics dealing with such conditions as arthritis, asthma, multiple sclerosis, lung and colon cancers and Alzheimer ft.s disease.
Specifications

Specifications

CERK
Polyclonal
PBS with 0.08% sodium azide
Q8TCT0
EC 2.7.1.138 antibody, Acylsphingosine kinase antibody, CERK antibody, Lipid kinase 4 antibody, LK4 antibody.
Rabbit
IgG
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
64781
ELISA, Western Blot
Unconjugated
CERK
Liquid
CERK
Synthetic peptide corresponding to human ceramide kinase.
Ammonium sulfate precipitation
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit