Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

COL4A3BP, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA112911

Catalog No. PA112911

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This gene encodes a kinase that specifically phosphorylates the N-terminal region of the non-collagenous domain of the alpha 3 chain of type IV collagen, known as the Goodpasture antigen. Goodpasture disease is the result of an autoimmune response directed at this antigen. One isoform of this protein is also involved in ceramide intracellular transport. Three transcript variants encoding different isoforms have been found for this gene.

A suggested positive control for this product is ovary lysate.
Specifications

Specifications

COL4A3BP
Polyclonal
PBS with 0.08% sodium azide
Q9Y5P4
CERT, CERTL, FLJ20597, GPBP, STARD11
Rabbit
IgG
100μg
-20° C, Avoid Freeze/Thaw Cycles, Avoid Freeze/Thaw Cycles
Primary
10087
Western Blot
Unconjugated
COL4A3BP
Liquid
COL4A3BP
Synthetic peptide corresponding to human CERTL protein.
Ammonium sulfate precipitation
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit