Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CES2 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA524157
Description
CES2 is a member of the carboxylesterase large family. The family members are responsible for the hydrolysis or transesterification of various xenobiotics, such as cocaine and heroin, and endogenous substrates with ester, thioester, or amide bonds. They may participate in fatty acyl and cholesterol ester metabolism, and may play a role in the blood-brain barrier system. The protein encoded by this gene is the major intestinal enzyme and functions in intestine drug clearance.Specifications
CES2 | |
Polyclonal | |
CES2 | |
CE-2, CES2A1, PCE-2, iCE, carboxylesterase 2 (intestine, liver), cocaine esterase, hCE-2, intestinal carboxylesterase; liver carboxylesterase-2, methylumbelliferyl-acetate deacetylase 2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
8824 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunohistochemistry, Western Blot | |
Unconjugated | |
O00748 | |
CES2 | |
KLH conjugated synthetic peptide between 340-369 amino acids from the central region of human CES2 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction