Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-CETP, Polyclonal, Novus Biologicals™

Goat Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP152162

Catalog No. NBP152162


Add to cart

Description

Description

CETP Polyclonal antibody specifically detects CETP in Human samples. It is validated for Peptide ELISA.
Specifications

Specifications

CETP
0.5 mg/mL
Peptide ELISA Detection limit 1:64000
NP_000069.2
cholesteryl ester transfer protein, cholesteryl ester transfer protein, plasma, HDLCQ10, Lipid transfer protein I
Goat
IgG
0.1 mg
Lipid and Metabolism
Primary
1071
ELISA
Unconjugated
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Affinity Purified
CETP
Peptide with sequence GNAHACSKGTSHEA corresponding to N-Terminus according to NP_000069.2.
Immunogen affinity purified
RUO
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only