Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CETP Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA514341

Catalog No. PIPA514341

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Cholesteryl ester transfer protein (CETP) transfers cholesteryl esters between lipoproteins. CETP may effect susceptibility to atherosclerosis.
Specifications

Specifications

CETP
Polyclonal
PBS with 0.09% sodium azide
P11597
Lipid transfer protein I; Cholesteryl ester transfer protein
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
1071
Western Blot
Unconjugated
CETP
Liquid
CETP
KLH conjugated synthetic peptide between 34-65 amino acids from the N-terminal region of human CETP
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit