Learn More
Anti- Cetuximab (Anti-Idiotype) Alexa Fluor™ 594-conjugated Antibody, R&D Systems™
Rabbit Monoclonal Antibody
Supplier:  R&D Systems FAB9626T100UG
Catalog No. FAB9626T100
Explore available promotions
Add to Cart
This item is not returnable. View return policy
Description
Description
Cetuximab Monoclonal specifically detects Cetuximab in Multi-Species samples. It is validated for Flow Cytometry.
Specifications
Specifications
Cetuximab
Monoclonal
Alexa Fluor 594
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Full length Cetuximab,
100 μg
Primary
Multi-species
Purified
Flow Cytometry
2259C
Flow Cytometry 0.25-1 ug/10^6 cells
Rabbit
Protein A or G purified from cell culture supernatant
RUO
Detects Cetuximab in Western blots and direct ELISAs. No cross-reactivity with Adalimumab, Rituximab, Trastuzumab, Brentuximab, Gemtuzumab, Ibritumomab, Tositumomab, Solanezumab, Teprotumumab, and Aducanumab is observed.
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
IgG
Product Suggestions
Product Suggestions