Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C6orf62 Rabbit anti-Hamster, Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524477

Catalog No. PIPA524477

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

C6orf62
Polyclonal
PBS with 0.09% sodium azide
Q9GZU0, Q99LU8
Nbla00237, XTP12, dJ30M3.2, HBV X-transactivated gene 12 protein, HBV X-transactivated protein 12, HBV XAg-transactivated protein 12, uncharacterized protein C6orf62
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
79555, 81688
Immunohistochemistry, Western Blot
Unconjugated
C6orf62
Liquid
BC005537, C6orf62
KLH conjugated synthetic peptide between 118-147 amino acids from the central region of human CF062
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Hamster, Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit