Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C6orf97 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526668

Catalog No. PIPA526668


Add to cart

Specifications

Specifications

C6orf97
Polyclonal
PBS with 0.09% sodium azide
Q8IYT3
bA282P11.1, coiled-coil domain-containing protein C6orf97
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
80129
Western Blot
Unconjugated
CCDC170
Liquid
CCDC170
KLH conjugated synthetic peptide between 594-623 amino acids from the C-terminal region of human CF097
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit