Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RRP36 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524273

Catalog No. PIPA524273

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with bovine based on sequence homology.

RRP36 functions at an early stage in the processing of 35S preribosomal RNA into the mature 18S species.
Specifications

Specifications

RRP36
Polyclonal
PBS with 0.09% sodium azide
Q96EU6, Q3UFY0
C6orf153, dJ20C7.4, RNA processing factor, ribosomal RNA processing protein 36 homolog
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
224823, 88745
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
RRP36
Liquid
RRP36
KLH conjugated synthetic peptide between 230-259 amino acids from the C-terminal region of human CF153
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit