Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Complement Factor B Monoclonal Antibody (313011), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA524036
Description
This antibody does not cross-react with recombinant human (rh) Factor D or rhFactor I. Reconstitute at 0.5 mg/mL in sterile PBS.
Complement Factor B Monoclonal antibody specifically detects Complement Factor B in Human samples. It is validated for Immunoprecipitation, Western Blot.Specifications
Complement Factor B | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P00751 | |
CFB | |
Mouse myeloma cell line NS0-derived recombinant human Complement Factor B Thr26-Leu764 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
Immunoprecipitation, Western Blot | |
313011 | |
Unconjugated | |
CFB | |
AHUS4; AI195813; AI255840; alternative-complement pathway C3/C5 convertase; ARMD14; B; BF; B-factor, properdin; BFD; C2; C3 proaccelerator; C3 proactivator; C3/C5 convertase; CFAB; CFB; CFBD; complement component 2 (within H-2S); complement factor B; Complement factor B Ba fragment; Complement factor B Bb fragment; Da1-24; DADB-122G4.5; Factor B; Fb; FBI12; GBG; glycine-rich beta glycoprotein; glycine-rich beta-glycoprotein; H2-Bf; histocompatibility 2, complement component factor B; PBF2; properdin; properdin factor B | |
Mouse | |
Protein A/G | |
RUO | |
629 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction