Learn More
Complement Factor H Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA524425
Catalog No. PIPA524425
Add to cart
Description
Description
This gene is a member of the Regulator of Complement Activation (RCA) gene cluster and encodes a protein with twenty short concensus repeat (SCR) domains. This protein is secreted into the bloodstream and has an essential role in the regulation of complement activation, restricting this innate defense mechanism to microbial infections. Mutations in this gene have been associated with hemolytic-uremic syndrome (HUS) and chronic hypocomplementemic nephropathy. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
Specifications
Specifications
Complement Factor H
Polyclonal
Unconjugated
CFH
Liquid
CFH
KLH conjugated synthetic peptide between 751-780 amino acids from the central region of human CFH
Protein A, Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide
P08603
AHUS1, AMBP1, ARMD4, ARMS1, CFHL3, FH, FHL1, HF, HF1, HF2, HUS, H factor 1 (complement), H factor 2 (complement), adrenomedullin binding protein, age-related maculopathy susceptibility 1, beta-1-H-globulin, beta-1H, factor H, factor H-like 1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
3075