Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Properdin Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA526939
Description
Properdin Polyclonal antibody specifically detects Properdin in Human samples. It is validated for Flow Cytometry, Western Blot.Specifications
Properdin | |
Polyclonal | |
Unconjugated | |
CFP | |
BCFG; Bf; BFD; C3 Proaccelerator; C3/C5 convertase; cfb; CFP; Complement factor P; complement factor properdin; GBG; H2-Bf; PBF2; PFC; PFD; PROPERDIN; properdin factor, complement; properdin P factor, complement | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
5199 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Western Blot | |
0.28 mg/mL | |
PBS with 0.09% sodium azide | |
P27918 | |
CFP | |
KLH conjugated synthetic peptide between 197-223 amino acids from the central region of human CFP | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction