Learn More
Mycobacterium tuberculosis CFP-10 Rabbit anti-Bacteria, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA119445
Catalog No. 01-676-760
Add to cart
Description
Description
This antibody recognizes CFP10 of Mycobacterium tuberculosis H37Rv and Mycobacterium bovis; it reacts also with recombit antigen produced in Escherichia coli.
Tuberculosis is a chronic bacterial infection which remains one of the leading killers of young adults world wide. Mycobacterium tuberculosis enters the alveoli by airborne transmission. They are engulfed by alveolar macrophages but resist destruction. Although the exact mechanism is not known, it is suspected that that Mycrobacterium tuberculosis survives by preventing the fusion of the phagosome and lysosome. As the bacterium accumulates it forms a tubercle, spreads to the lymph nodes and becomes a systemic infection. Although treatments for tuberculosis do exist, new strains of Mycobacterium tuberculosis that appear to have multiple-drug resistance (MDR) pose a considerable threat to public health.
Specifications
Specifications
Mycobacterium tuberculosis CFP-10
Polyclonal
Unconjugated
Liquid
Rabbit
IgG
100 μg
4° C, do not freeze
Primary
Bacteria
Western Blot
1 mg/mL
PBS with 15mM sodium azide; pH 7.4
M. tuberculosis; myobacter
CFP10 (Rv3874) of Mycobacterium tuberculosis.
Affinity chromatography
RUO
Antibody
Polyclonal