Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

glycoprotein hormones, alpha polypeptide, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human CGA protein.

Manufacturer:  Abnova Corporation H00001081B01

Catalog No. 89-005-122


Add to cart

Description

Description

The four human glycoprotein hormones chorionic gonadotropin (CG), luteinizing hormone (LH), follicle stimulating hormone (FSH), and thyroid stimulating hormone (TSH) are dimers consisting of alpha and beta subunits that are associated noncovalently. The alpha subunits of these hormones are identical, however, their beta chains are unique and confer biological specificity. The protein encoded by this gene is the alpha subunit and belongs to the glycoprotein hormones alpha chain family. [provided by RefSeq

Sequence: MDYYRKYAAIFLVTLSVFLHVLHSAPDVQDCPECTLQENPLFSQPGAPILQCMGCCFSRAYPTPLRSKKTMLVQKNVTSESTCCVAKSYNRVTVMGGFKVENHTACHCSTCYYHKS
Specifications

Specifications

glycoprotein hormones, alpha polypeptide
Unconjugated
No additive
BC010957
CG-ALPHA|FSHA|GPHA1|GPHa|HCG|LHA|TSHA
Mouse
50 uL
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
1081
Western Blot
Mouse polyclonal antibody raised against a full-length human CGA protein.
CGA
Serum
CGA
CGA (AAH10957, 1 a.a. ~ 116 a.a) full-length human protein.
RUO
Y
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit