Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cGB-PDE Mouse anti-Rat, Unlabeled, Clone: 54, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 611499

 View more versions of this product

Catalog No. BDB611499


Add to cart

Description

Description

cGMP is involved in nitric oxide signaling, as well as cell signaling associated with natriuretic peptides and guanylins. Intracellular targets for cGMP include cGMP-dependent protein kinase, cyclic nucleotide-gate ion channels, and cGMP-Binding PhosphoDiesterases (cGB-PDE). Tissue levels of cGMP are determined by guanylyl cyclases that catalyze the formation of cGMP and cyclic nucleotide PDEs that catalyze the breakdown of cGMP. In the PDE superfamily PDE2, PDE5, PDE6, and PDE10 have dimeric structures that contain highly cGMP-specific allosteric cGMP binding sites in addition to a phosphodiesterase catalytic site. PDE5 is highly expressed in aorta and lung, but is also expressed in intestine, kidney, adrenal gland, cerebellum, and cerebrum. In the cerebellum, PDE5 is expressed in the Purkinje cell layer during neonatal development. PDE5 is also abundant in vascular smooth muscle cells where it is involved in regulating cGMP levels and vascular smooth muscle tonicity. Thus, PDE5 is important for regulating cGMP levels in both neuronal and non-neuronal tissues that utilize cGMP signaling pathways for the regulation of cell function.

Host Species: Mouse
Clone: 54
Isotype: IgG1
Species Reactivity: Rat
Immunogen: Rat cGB-PDE aa. 2-108
Formula Weight [Chemical]: 85-95kDa

Immunofluorescence, Western Blotting

Specifications

Specifications

cGB-PDE
54
Unconjugated
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Western Blot
250μg/mL
PDE5; cGMP-Binding PhosphoDiesterases
Affinity Purified
Rat cGB-PDE aa. 2-108
150μg
Cell Biology
Primary
Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.