Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CGRP Guinea Pig anti-Human, Rat, Polyclonal, Invitrogen™

Guinea Pig Polyclonal Antibody

Manufacturer:  Invitrogen PA136017

Catalog No. 01-672-207


Add to cart

Description

Description

This antibody cross reacts with amylin (IAPP). For immunofluorescence applications, overnight incubation at 4°C is required. Stefanini fixed frozen sections of rat colon can be used as a positive control for this antibody. Reconstitute with 50-100 µL of distilled water, and dilute further in 0.1M PBS with 1% BSA and 0.1% sodium azide. Once reconstituted, store at -20°C.

Calcitonin is a peptide hormone synthesized by the parafollicular cells of the thyroid. CGRP is one of the most abundant peptides produced in both peripheral and central neurons. It is a potent peptide vasodilator and can function in the transmission of pain.
Specifications

Specifications

CGRP
Polyclonal
whole serum with 0.1% sodium azide
P01258, P01256
Calca, CA
Guinea Pig
IgG
RUO
Antibody
Polyclonal
Human, Rat
Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Unconjugated
CALCA
Lyophilized
CALCA
Synthetic human CGRP.
50 μL
4° C
Primary
24241, 796
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit