Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CGRP Guinea Pig anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Guinea Pig Polyclonal Antibody

Manufacturer:  Invitrogen PA136018

Catalog No. 01-672-208


Add to cart

Description

Description

Reconstitute with 50-100 µL of distilled water, and dilute further in 0.1M PBS with 1% BSA and 0.1% sodium azide. For long-term storage, aliquot and store at -20°C.

This gene encodes the peptide hormones calcitonin, calcitonin gene-related peptide and katacalcin by tissue-specific alternative RNA splicing of the gene transcripts and cleavage of inactive precursor proteins. Calcitonin is involved in calcium regulation and acts to regulate phosphorus metabolism. Calcitonin gene-related peptide functions as a vasodilator while katacalcin is a calcium-lowering peptide. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications

Specifications

CGRP
Polyclonal
whole serum with 0.1% sodium azide
P01258, P70160, P01256
Calca, CA
Guinea Pig
IgG
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Unconjugated
CALCA
Lyophilized
CALCA
Synthetic rat CGRP conjugated to BSA.
50 μL
4° C
Primary
12310, 24241, 796
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit