Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CGRP Rabbit anti-Human, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA136049

Catalog No. 01-672-235


Add to cart

Description

Description

Reconstitute with 50-100 µL of distilled water, and dilute further in 0.1M PBS with 1% BSA and 0.1% sodium azide.

Calcitonin is a peptide hormone synthesized by the parafollicular cells of the thyroid. CGRP is one of the most abundant peptides produced in both peripheral and central neurons. It is a potent peptide vasodilator and can function in the transmission of pain.
Specifications

Specifications

CGRP
Polyclonal
whole serum with 0.1% sodium azide
P01258, P01256
Calca, CA
Rabbit
IgG
RUO
Antibody
Polyclonal
Human, Rat
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Unconjugated
CALCA
Lyophilized
CALCA
Synthetic porcine CGRP conjugated to BSA.
50 μL
4° C
Primary
24241, 796
Documents
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit