Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ch-TOG Rabbit anti-Human, Mouse, Amphibian, Xenopus, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Supplier: Novus Biologicals NBP106513
Description
ch-TOG Polyclonal specifically detects ch-TOG in Human, Mouse, Amphibian, Xenopus samples. It is validated for Western Blot, Immunoprecipitation.Specifications
ch-TOG | |
Polyclonal | |
Unconjugated | |
TBS and 0.1% BSA with 0.09% Sodium Azide | |
Ch-TOG, CHTOG, Colonic and hepatic tumor over-expressed gene protein, cytoskeleton associated protein 5, cytoskeleton-associated protein 5, EC 2.7.7.6, EC 3.1.1.29, KIAA0097FLJ35359, MSPS, TOG, TOGp | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse. Based on 100% sequence identity, this antibody is predicted to react with Rat, X. laevis, X. tropicalis, Chicken, Turkey, Bovine, Dog, Horse, Rabbit, Guinea pig_10141, Pig, Panda, Orangutan, Rhesus Monkey, Gorilla, Chimpanzee, White-tufted-ear marmoset, Chinese hamster, and Naked mole rat. | |
Store at 4C. Do not freeze. |
Western Blot, Immunoprecipitation | |
0.2 mg/mL | |
Western Blot 1:2000-1:10000, Immunoprecipitation 2-5 ug/mg lysate | |
Q14008 | |
CKAP5 | |
The immunogen recognized by this antibody maps to a region between residue 1982 and 2032 of human colonic and hepatic tumor over-expressed protein using the numbering given in entry NP_001008938.1 (GeneID 9793). | |
0.1 mL | |
Cell Cycle and Replication | |
9793 | |
Human, Mouse, Amphibian, Xenopus | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only