Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSP60 Rabbit anti-Bovine, Canine, Chicken, Human, Non-human primate, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA141662

Catalog No. 01-675-271


Add to cart

Description

Description

Suggested positive control: HeLa, Jurkat cell lysate, Hela whole cell lysate, antigen standard for HSPD1 (transient overexpression lysate), Jurkat whole cell lysate.

HSP60, a 60KDa member of Chaperonin family, is a mitochondrial protein that participates in protein folding, transport and secretion. HSP60 is a multi-functional protein whose roles include accelerating the maturation of pro-caspase 3 during apoptosis, LPS-binding, acting as a ligand for innate immune system and also binding to the surface of innate immune cells and stimulating proinflammatory reactivity. It induces the expression of IL-12 and IL-15 and also induces release of TNF-alpha, NO and IL-6. Reports suggest interaction of Hsp60 with integrin molecules during metastasis. Mutations in Hsp60 gene cause autosomal recessive spastic paraplegia 13. Hsp60 has been found to be involved in development of auto-immune disorders such as Rheumatoid arthritis, insulin-dependent diabetes mellitus, systemic sclerosis and atherosclerosis.
Specifications

Specifications

HSP60
Polyclonal
Unconjugated
HSPD1
Liquid
HSPD1
Synthetic peptide corresponding to residues 350-400 of human Hsp60.
Protein G
RUO
Antibody
Polyclonal
Bovine, Canine, Chicken, Human, Non-human Primate
Western Blot
0.5 mg/mL
PBS with 0.2% gelatin and 0.05% sodium azide
P31081, P49818, Q5ZL72, P10809
CPN60; GROEL; Mytonin Chaperone 60; HLD4; HSP-60; HSP60; HSP65; HuCHA60; SPG13; 60 kDa chaperonin; 60 kDa heat shock protein; mitochondrial; P60 lymphocyte protein; chaperonin 60; heat shock protein 65; mitochondrial matrix protein P1; short heat shock protein 60 Hsp60s1
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
3329, 424059, 478854, 511913
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit