Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ChAT Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526597

Catalog No. PIPA526597


Add to cart

Description

Description

CHAT is an enzyme which catalyzes the biosynthesis of the neurotransmitter acetylcholine. This gene product is a characteristic feature of cholinergic neurons, and changes in these neurons may explain some of the symptoms of Alzheimer disease.
Specifications

Specifications

CHAT
Polyclonal
PBS with 0.09% sodium azide
P28329, Q03059
CHOACTASE, CMS1A, CMS1A2, acetyl CoA:choline O-acetyltransferase, choline acetylase
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
1103, 12647
Immunofluorescence, Western Blot
Unconjugated
CHAT
Liquid
CHAT
KLH conjugated synthetic peptide between 98-128 amino acids from the N-terminal region of human CHAT
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit