Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CHD1 Monoclonal Antibody (677616), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA524306
Description
In direct ELISAs, no cross-reactivity with recombinant human CHD1L, 5, or 7 is observed. Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.
CHD1 Monoclonal antibody specifically detects CHD1 in Human samples. It is validated for Immunocytochemistry, Western Blot.Specifications
CHD1 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
O14646 | |
CHD1 | |
E. coli-derived recombinant human CHD1 Lys1531-Thr1710 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunocytochemistry, Western Blot | |
677616 | |
Unconjugated | |
CHD1 | |
4930525N21Rik; AI851787; ATP-dependent helicase CHD1; AW555109; Chd1; Chd-1; chromodomain helicase DNA binding protein 1; chromodomain-helicase-DNA-binding protein 1; DKFZp686E2337; EC 3.6.1; EC 3.6.4.12; LOW QUALITY PROTEIN: chromodomain-helicase-DNA-binding protein 1 | |
Mouse | |
Protein A/G | |
RUO | |
1105 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction