Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

chromodomain helicase DNA binding protein 2, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human CHD2 protein.

Manufacturer:  Abnova Corporation H00001106B01

Catalog No. 89-005-125


Add to cart

Description

Description

The CHD family of proteins is characterized by the presence of chromo (chromatin organization modifier) domains and SNF2-related helicase/ATPase domains. CHD genes alter gene expression possibly by modification of chromatin structure thus altering access of the transcriptional apparatus to its chromosomal DNA template. Alternatively spliced transcript variants encoding distinct isoforms have been found for this gene. [provided by RefSeq

Sequence: MMRNKDKSQEEDSSLHSNASSHSASEEASGSDSGSQSESEQGSDPGSGHGSESNSSSESSESQSESESESAGSKSQPVLPEAKEKPASKKERIADVKKMWEEYPDVYGVRRSNRSRQEPSRFNIKEEASSGSESGSPKRRGQRQLKKQEKWKQEPSEDEQEQGTSAESEPEQKKVKARRPVPRRTVPKPRVKKQPKTQRGKRKKQDSSDEDDDDDEAPKRQTRRRAAKNVSYKEDDDFETDSDDLIEMTGEGVDEQQDNSETIEKVLDSRLGKKGATGASTTVYAIEANGDPSGDFDTEKDEGEIQYLIKWKGWSYIHSTWESEESLQQQKVKGLKKLENFKKKEDEIKQWLGKVSPEDVEYFNCQQELASELNKQYQIVERVIAVKTSKSTLGQTDFPAHSRKPAPSNEPEYLCKWMGLPYSECSWEDEALIGKKFQNCIDSFHSRNNSKTIPTRECKALKQRPRFVALKKQPAYLGGENLELRDYQLEGLNWLAHSWCK
Specifications

Specifications

chromodomain helicase DNA binding protein 2
Unconjugated
No additive
NM_001042572.1
DKFZp547I1315|DKFZp686E01200|DKFZp781D1727|FLJ38614
Mouse
50 uL
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
1106
Immunofluorescence, Western Blot
Mouse polyclonal antibody raised against a full-length human CHD2 protein.
CHD2
Serum
CHD2
CHD2 (AAH07347.1, 1 a.a. ~ 501 a.a) full-length human protein.
RUO
Y
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit