Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHD3 Mouse, Unlabeled, Clone: 44, BD

Mouse Monoclonal Antibody

Specifications

Antigen CHD3
Isotype IgG1
Immunogen Human CHD3 aa. 1277-1474
Conjugate Unconjugated
Primary or Secondary Primary
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
BDB611846
View Documents Promotion Details
BD Biosciences
611846
50μg This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
BDB611847
View Documents Promotion Details
BD Biosciences
611847
150μg This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

The SNF2-related family of proteins all contain conserved helicase motifs, and have been implicated in transcription regulation, DNA repair, and DNA recombination. The DNA-dependent ATPase activity of these proteins is usually activated by single-stranded, double-stranded, or nucleosomal DNA. The chromodomain helicase DNA binding proteins (CHDs) are a subfamily of the SNF2 family, and includes CHD1, CHD2, CHD3, and CHD4. CHD3 and CHD4 contain N-terminal paired PHD zinc finger motifs (PZF), two heterochromatin binding domains (chromodomains; CDs), and a central SNF2-like helicase/ATPase domain. CHD3 mRNA is widely expressed, and CHD3 protein localizes to the nucleus, but not nucleoli. Both CHD3 and CHD4 were first identified as self-antigens recognized by sera from patients with the autoimmune connective-tissue disease Mi-2 dermatomyositis. In addition, both CHD3 and CHD4 are associated with a nucleosome remodelling and deacetylating (NRD) complex, and facilitate deacetylation of oligonucleosomal histones in vitro. Thus, CHD3 and CHD4 are chromodomain containing proteins that may have important roles in ATP-dependent nucleosome remodelling.

Host Species: Mouse
Clone: 44
Isotype: IgG1
Species Reactivity [for Features Main]: Human
Immunogen: Human CHD3 aa. 1277-1474

Immunofluorescence, Western Blotting

Specifications

Specifications

CHD3
Human CHD3 aa. 1277-1474
Primary
Immunofluorescence, Western Blot
RUO
44
250μg/mL
IgG1
Unconjugated
Cell Biology
Murine
Affinity Purified
Monoclonal
SDS
Product Certifications

For Research Use Only.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit