Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chemerin Mouse anti-Human, Clone: 365314, R&D Systems™

Mouse Monoclonal Antibody

$119.60 - $839.00

Specifications

Antigen Chemerin
Immunogen E. coli-derived recombinant human Chemerin Glu21-Ser157 Accession # Q99969
Clone 365314
Applications ELISA
Conjugate Unconjugated
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB23241SP
View Documents
R&D Systems
MAB23241SP
25 ug
Each for $119.60
Add to cart
 
MAB23241100
View Documents
R&D Systems
MAB23241100
100 ug
Each for $377.00
Add to cart
 
MAB23241500
View Documents
R&D Systems
MAB23241500
500 ug
Each for $839.00
Add to cart
 
Description

Description

Chemerin Monoclonal antibody MAB23241 Specifically detects Chemerin in Human samples. It is validated for ELISA Capture
Specifications

Specifications

Chemerin
365314
Unconjugated
Q99969
Monoclonal
Primary
Detects human Chemerin in ELISAs. In ELISAs, this antibody does not cross-react with recombinant mouse Chemerin.
Protein A or G purified from hybridoma culture supernatant
E. coli-derived recombinant human Chemerin Glu21-Ser157 Accession # Q99969
ELISA
Purified
Mouse
IgG2b
5919
RUO
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit